  • Kritériumok
    • Raktáron (5)
  • Gyártó
  • Szűrés gyártók szerint
    Rendezés:
Ön erre keresett rá:
Sejtkaparók
17  results were found
https://hu.vwr.com/stibo/smallweb/std.lang.all/96/43/18709643.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"17"
  Lista nézet Easy View (new)
Rendezés:
Gyártó:  BÜRKLE
Leírás:   PS, white. Sturdy and convenient scrapers for disposable use. The scrapers are suitable for scraping off, smoothing or cleaning as well as removing re...
Katalógus szám: (BURK5378-2030)
Gyártó:  BÜRKLE
Leírás:   PS, blue. This scraper is not only coloured blue but also has a special additive in the material that makes it visible to metal detectors or X rays. T...
UOM:  1 * 10 DB
Gyártó:  Bohlender
Leírás:   Scrapers made of PTFE with tapered end.
Katalógus szám: (BURK5378-3030)
Gyártó:  BÜRKLE
Leírás:   PS, blue. This scraper is suitable for scraping off, smoothing or cleaning as well as removing residue. Especially designed for use in food, animal fe...
UOM:  1 * 10 DB
Gyártó:  Sampling Systems
Leírás:   These disposable scrapers are made from high quality HDPE. They are ideal in the manufacture of creams, ointments and pastes.
Gyártó:  BÜRKLE
Leírás:   These scrapers made of green PP are ideal for industrial applications, such as pharmaceuticals, biotechnology, foods, chemistry and other industries.
Gyártó:  BÜRKLE
Leírás:   These scrapers made of white PP are ideal for use with foodstuffs.
Gyártó:  MICRONOVA
Leírás:   PP, white.
1 - 8  of 17
  1  2  3  Következő