  • Kritériumok
    • Raktáron (2)
  • Gyártó
  • Szűrés gyártók szerint
    Rendezés:
Ön erre keresett rá:
Porszívók
27  results were found
https://hu.vwr.com/stibo/smallweb/std.lang.all/69/58/20236958.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"27"
  Lista nézet Easy View (new)
Rendezés:
Katalógus szám: (181-0341)
Gyártó:  TIGER-VAC
Leírás:   [EN]VACUUM MODEL EMI-1000 240V 1 * 1 db
UOM:  1 * 1 db
Gyártó:  TIGER-VAC
Leírás:   Industrial vacuum cleaner for continuous duty. Designed specifically for the food and pharmaceutical industries. It is ideal for general cleaning and ...
UOM:  1 * 1 DB
Gyártó:  TIGER-VAC
Leírás:   Designed for general clean up in contamination controlled environments, cleanroom compatible and ideal for pharmaceutical industry.
Katalógus szám: (TIGV111413B)
Gyártó:  TIGER-VAC
Leírás:   This industrial vacuum cleaner is designed for the food industry.
UOM:  1 * 1 DB
Katalógus szám: (141-1212)
Gyártó:  TIGER-VAC
Leírás:   This wet and dry recovery vacuum cleaner is cleanroom compatible and can be used in pharmaceutical industry. It has been designed for the recovery of ...
UOM:  1 * 1 db
Katalógus szám: (TIGV110540B)
Gyártó:  TIGER-VAC
Leírás:   Wet and dry recovery cleaner designed for the recovery of broken ampules and glass in contamination controlled environments. The broken ampules and gl...
UOM:  1 * 1 DB
Gyártó:  TIGER-VAC
Leírás:   This vacuum cleaner is equipped with a backpack harness, it is lightweight and ergonomic. It is designed for general cleanup. The construction is all ...
UOM:  1 * 1 DB
Katalógus szám: (TIGV110750B)
Gyártó:  TIGER-VAC
Leírás:   This vaccum cleaner made of heavy duty plastic, is designed for the recovery of lead, asbestos, silica and other toxic/nuisance dusts.
UOM:  1 * 1 DB
1 - 8  of 27
  1  2  3  4  Következő