Folytatni kívánja a munkamenetet?
Kilépés 
A munkamenet lejárt
A munkamenet lejárt. Biztonsági okokból kijelentkeztettük.
Szeretne újra bejelentkezni?

Anti-MAOB Goat Polyclonal Antibody

Gyártó: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 60 kDa band observed in human brain lysates (calculated MW of 58.8 kDa according to NP_000889.3). Recommended concentration: 1-3 μg/mL. an additional band of unknown identity was also consistently observed at 150 kDa . This band was successfully blocked by incubation with the immunizing peptide.

MAOB antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MAOB antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

RENDELÉS

Többet mutat / Kevesebbet mutat
Méret M.egys VWR katalógus szám Mennyiség
100 µg 100 µG PRSI45-862
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása

SPECIFIKÁCIÓK

Antigen symbol MAOB
Antigen name Monoamine Oxidase B
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
Antigen synonyms amine oxidase (flavin-containing), tyramine oxidase, adrenalin oxidase, platelet, HGNC:6834, MGC26382, MAO, monoamine oxidase B , MAOB, brain, RP1-201D17__B.1
Koncentráció 500 ug/mL
Immunogen MAOB antibody was raised against a 13 amino acid synthetic peptide near the internal region of MAOB.
Form Liquid
Adja meg a részleteket

Válassza ki a cikket, amihez tanúsítványt keres.

Kérjük, adja meg a sarzs számot! A mező kitöltése kötelező, ismeretlen szám esetén használjon x-et.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Vagy
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség