|
![]() |
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | M6PR |
Antigen name | Mannose-6-Phosphate Receptor |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Host | Goat |
Western blot | Yes |
ELISA | Yes |
Antigen synonyms | Cation-dependent mannose-6-phosphate receptor, CD-MPR, 46 kDa mannose 6-phosphate receptor, MPR46, CD Man-6-P receptor, M6PR, MPR 46, MPRD |
Koncentráció | 500 ug/mL |
Immunogen | M6PR antibody was raised against a 14 amino acid synthetic peptide near the internal region of M6PR. |
Form | Liquid |