Folytatni kívánja a munkamenetet?
Kilépés 
A munkamenet lejárt
A munkamenet lejárt. Biztonsági okokból kijelentkeztettük.
Szeretne újra bejelentkezni?

Anti-NR1H2 Goat Polyclonal Antibody

Gyártó: ProSci Inc.
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Experiments gave a band at approximately 80 kDa and a weak band at appr. 60 kDa in human bone marrow lysate after 1 μg/mL antibody staining. These bands correspond to earlier findings with different antibodies (PMID: 12393874-60 kDa and other unpublished antibodies-80 kDa ). This protein has a calculated MW of 51.0 kDa according to NP_009052. Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human spleen shows nucleolar staining. Recommended concentration, 1-2 μg/mL.

LXR antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

LXR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NR1H2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

RENDELÉS

Többet mutat / Kevesebbet mutat
Méret M.egys VWR katalógus szám Mennyiség
100 µg 100 µG PRSI45-849
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása

SPECIFIKÁCIÓK

Antigen symbol NR1H2
Antigen name Nuclear Receptor Subfamily 1, Group H, Member 2
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms UNR, LXR-b, RIP15, NER-I, NER, LXRB
Koncentráció 500 ug/mL
Immunogen LXR antibody was raised against a 15 amino acid synthetic peptide near the N-Terminus of LXR.
Form Liquid
Adja meg a részleteket

Válassza ki a cikket, amihez tanúsítványt keres.

Kérjük, adja meg a sarzs számot! A mező kitöltése kötelező, ismeretlen szám esetén használjon x-et.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Vagy
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség