Folytatni kívánja a munkamenetet?
Kilépés 
A munkamenet lejárt
A munkamenet lejárt. Biztonsági okokból kijelentkeztettük.
Szeretne újra bejelentkezni?

Anti-LRP5 Goat Polyclonal Antibody

Gyártó: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26 kDa band in lysates of human brain, of T cell line MOLT4, of myeloid erythroblast cell line K562 and of EBV-immortilized lymphoblastoid after 1 μg/mL antibody staining.1. The 26 kDa band was successfully blocked by incubation with the immunizing peptide. Also,. Western Blot: LRP5 antibody has been successfully used in WB assays with rabbit tissue lysate (whole bone; amount loaded: 20 μg; antibody dilution 1:200; incubation time of primary antibody: 2 hours; milk (5%) as blocking agent for 1.5 hours; sewcondary antibody: bovine anti-goat with incubation time of 2 hours). Rabbit Bone samples were used and probed at the same time with GAPDH as a loading control. Specific bands of 51 kDa (LRP5) and 37 kDa (GAPDH) were detected. Antibody buffer was 1% NFDM in TBS. (Information was provided by a customer). Immunohistochemistry: In paraffin embedded human pancreas shows staining of cytoplasm in some cells of the islets of Langerhans. Recommended concentration, 1-2 μg/mL.

LRP5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

LRP5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LRP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

RENDELÉS

Többet mutat / Kevesebbet mutat
Méret M.egys VWR katalógus szám Mennyiség
100 µg 100 µG PRSI45-846
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása

SPECIFIKÁCIÓK

Antigen symbol LRP5
Antigen name Low Density Lipoprotein Receptor-related Protein 5
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms LR3, LRP-5, OPS, BMND1, OPTA1, HBM, EVR4, VBCH2, EVR1, LRP7, OPPG
Koncentráció 500 ug/mL
Immunogen LRP5 antibody was raised against a 12 amino acid synthetic peptide near the internal region of LRP5.
Form Liquid
Adja meg a részleteket

Válassza ki a cikket, amihez tanúsítványt keres.

Kérjük, adja meg a sarzs számot! A mező kitöltése kötelező, ismeretlen szám esetén használjon x-et.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Vagy
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség