Folytatni kívánja a munkamenetet?
Kilépés 
A munkamenet lejárt
A munkamenet lejárt. Biztonsági okokból kijelentkeztettük.
Szeretne újra bejelentkezni?

Anti-XRCC6 Goat Polyclonal Antibody

Gyártó: ProSci Inc.
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 70 kDa band observed in lysates of cell lines A431, HeLa and HepG2 (calculated MW of 69.8 kDa according to NP_001460.1). In transfected HEK293 transiently expressing KU70 a band of approximately 70 kDa is observed. This band is not observed in the non-transfected HEK293.Recommended concentration: 0.1-0.3 μg/mL.

KU70 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

KU70 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: XRCC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

RENDELÉS

Többet mutat / Kevesebbet mutat
Méret M.egys VWR katalógus szám Mennyiség
100 µg 100 µG PRSI45-817
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása

SPECIFIKÁCIÓK

Antigen symbol XRCC6
Antigen name X-ray repair complementing defective repair in Chinese hamster cells 6
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
Antigen synonyms CTCBF, ATP-dependent DNA helicase 2 subunit 1, KU70, 5'-deoxyribose-5-phosphate lyase Ku70, ATP-dependent DNA helicase II, 70 kDa subunit of Ku antigen, ATP-dependent DNA helicase II 70 kDa subunit, CTC box binding factor 75 kDa subunit, ML8, TLAA, G22P1, CT, 5'-dRP lyase Ku70, CTC75, 70 kDa subunit
Koncentráció 500 ug/mL
Immunogen KU70 antibody was raised against a 13 amino acid synthetic peptide near the N-Terminus of KU70.
Form Liquid
Adja meg a részleteket

Válassza ki a cikket, amihez tanúsítványt keres.

Kérjük, adja meg a sarzs számot! A mező kitöltése kötelező, ismeretlen szám esetén használjon x-et.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Vagy
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség