|
![]() |
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | KPNA3 |
Antigen name | Karyopherin Alpha 3 |
Reactivity | Human, Rat, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Host | Goat |
Western blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
ImmunoPrecipitation | Yes |
ImmunoFluorescence | Yes |
Antigen synonyms | KPNA3, hSRP1, SRP1, SRP4, Importin alpha Q2, QIP2, Importin subunit alpha-4, SRP1gamma, IPOA4, Qip2 |
Koncentráció | 500 ug/mL |
Immunogen | KPNA3 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of KPNA3. |
Form | Liquid |