|
![]() |
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | KIF5A |
Antigen name | Kinesin Family Member 5A |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Host | Goat |
ELISA | Yes |
Antigen synonyms | MY050, NKHC1, NKHC, Kinesin heavy chain isoform 5A, SPG10, KIF5, D12S1889, Kinesin heavy chain neuron-specific 1 |
Koncentráció | 500 ug/mL |
Immunogen | KIF5A antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of KIF5A. |
Form | Liquid |