Folytatni kívánja a munkamenetet?
Kilépés 
A munkamenet lejárt
A munkamenet lejárt. Biztonsági okokból kijelentkeztettük.
Szeretne újra bejelentkezni?

Anti-IDE Goat Polyclonal Antibody

Gyártó: ProSci Inc.
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 110 kDa band observed in mouse brain and rat brain lysates (calculated MW of 118 kDa according to NP_112419.2). Recommended concentration: 1-3 μg/mL. an additional band of unknown identity was also consistently observed at 30 kDa . This band was successfully blocked by incubation with the immunizing peptide.

Ide antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Ide antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IDE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat




 

RENDELÉS

Többet mutat / Kevesebbet mutat
Méret M.egys VWR katalógus szám Mennyiség
100 µg 100 µG PRSI45-760
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása

SPECIFIKÁCIÓK

Antigen symbol IDE
Antigen name Insulin Degrading Enzyme
Reactivity Rat, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
Antigen synonyms 4833415K22Rik, AA675336, insulin-degrading enzyme, insulinase, Ide, INSULYSIN, 1300012G03Rik, insulysin, AI507533
Koncentráció 500 ug/mL
Immunogen Ide antibody was raised against an 11 amino acid synthetic peptide near the internal region of Ide.
Form Liquid
Adja meg a részleteket

Válassza ki a cikket, amihez tanúsítványt keres.

Kérjük, adja meg a sarzs számot! A mező kitöltése kötelező, ismeretlen szám esetén használjon x-et.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Vagy
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség