Folytatni kívánja a munkamenetet?
Kilépés 
A munkamenet lejárt
A munkamenet lejárt. Biztonsági okokból kijelentkeztettük.
Szeretne újra bejelentkezni?

Anti-SETD2 Goat Polyclonal Antibody

Gyártó: ProSci Inc.
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 80 kDa band in human brain (cerebellum, hippocampus, frontal cortex) lysates after 0.3 μg/mL antibody staining. The 80 kDa band was successfully blocked by incubation with the immunizing peptide. Approximately 290 kDa band observed in lysates of mouse fybroblasts with a genetic construct encoding siRNA. The observed band is knocked down by two different siRNA molecules. Calculated MW of 231 kDa according to NP_054878.3).The Ponceau red demonstrates proper protein loading. Recommended concentration: 0.5-2 μg/mL.

HYPB antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

HYPB antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SETD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse




Kérjük jelentkezzen be a saját árak és készletadatok megtekintéséhez!
  Remember me on this device
 

RENDELÉS

Többet mutat / Kevesebbet mutat
Méret M.egys VWR katalógus szám Mennyiség
100 µg 100 µG PRSI45-757
Vonalkód generálása Barcode Label Format: 
PDF katalógus oldal generálása

SPECIFIKÁCIÓK

Antigen symbol SETD2
Antigen name SET Domain Containing 2
Reactivity Mouse
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
Western blot Yes
ELISA Yes
Antigen synonyms p231HBP, HYPB, KMT3A, huntingtin interacting protein 1, HIP-1, HIF-1, SET2, lysine N-methyltransferase 3A, HBP231, huntingtin-interacting protein B, huntingtin yeast partner B, histone-lysine N-methyltransferase SETD2
Koncentráció 500 ug/mL
Immunogen HYPB antibody was raised against a 12 amino acid synthetic peptide near the internal region of HYPB.
Form Liquid
Adja meg a részleteket

Válassza ki a cikket, amihez tanúsítványt keres.

Kérjük, adja meg a sarzs számot! A mező kitöltése kötelező, ismeretlen szám esetén használjon x-et.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Vagy
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség