Szeretne újra bejelentkezni?
Environmental/HazMat racks for small containers
Gyártó: CEMO GMBH
For storage of water-polluting substances in small containers up to 30 l capacity.
  • With general construction inspection approval no. Z-40.22-388
  • Each level can support up to 200 kg
Environmental/HazMat rack 13/20 is completely hot-dip galvanised and equipped with a 20 l leak-tight tray bottom welded in accordance with StawaR (German guidelines for the requirements
of steel container trays of capacity up to 1000 l) lowest storage level.
Environmental/HazMat rack 10/20 is completely galvanised and can be assembled without screws or tools. It has 5 shelf floors which can be positioned in mounting points that have 25 mm spacing and two 30 l, PE collection trays.
RENDELÉS
Többet mutat / Kevesebbet mutat
Leírás Szél. × mélys. × mag. Tömeg M.egys VWR katalógus szám Mennyiség
Environmental/HazMat rack 10/20 for small containers 1060×630×2000 mm 44 kg 1 CEMG8091
Expansion unit for Environmental/HazMat rack 10/20 1010×630×2000 mm 34 kg 1 CEMG8092
Environmental/HazMat rack 13/20, for small containers, storage height from ground 160/690/1170/1650 mm 1320×630×2000 mm 92 kg 1 CEMG7820
Expansion unit for Environmental/HazMat rack 13/20, storage height from ground 160/690/1170/1650 mm 1320×630×2000 mm 82 kg 1 CEMG7856
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
ACCESSORIES
Többet mutat / Kevesebbet mutat
Accessories
Leírás Dla M.egys VWR katalógus szám Mennyiség
Additional shelf floor, 1000×600×40 mm, 4 kg Environmental/HazMat rack 10/20 1 CEMG8093
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség