  • Kritériumok
    • Raktáron (2)
  • Gyártó
  • Szűrés gyártók szerint
    Rendezés:
  • Keresés szűkítése
Védősisakok
https://hu.vwr.com/stibo/smallweb/std.lang.all/68/43/20236843.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"14"
Rendezés:
Gyártó:  SCOTT SAFETY
Leírás:   Bumpmaster is a lightweight and wipe clean low profile PE bump cap.
Gyártó:  SCOTT SAFETY
Leírás:   This stylish bump cap is made from lightweight, microfibre fabric. Its unique shell design delivers optimum protection and is shaped over the ear to p...
Katalógus szám: (111-1913)
Gyártó:  UVEX
Leírás:   This lightweight bump cap offers maximum wearer comfort and protection against bumps and grazes. The comfortable interior consists of four layers: Sho...
UOM:  1 * 1 DB
Katalógus szám: (SCTS2021191)
Gyártó:  SCOTT SAFETY
Leírás:   First Base 3 Beanie is made from elasticated knitted acrylic and provides a universal fit to a wide range of head sizes.
UOM:  1 * 1 DB
Gyártó:  SCOTT SAFETY
Leírás:   The safety baseball cap First Base Plus is made from durable ABS plastic with flexible shell design to protect the wearer from bumps and lacerations t...
Gyártó:  SCOTT SAFETY
Leírás:   This stylish bump cap is made from lightweight, microfibre fabric. Its unique shell design delivers optimum protection and is shaped over the ear to p...
Gyártó:  SCOTT SAFETY
Leírás:   First Base 3 Winter helps ensure workers stay warm and dry in cold and wet weather conditions, offering a practical indoor and outdoor bump protection...
Katalógus szám: (111-2260)
Gyártó:  SCOTT SAFETY
Leírás:   The First Base 3 flame-retardant bump protection cap is designed to protect workers where they are exposed to flammable liquids or hot sparks that cou...
UOM:  1 * 1 DB
1 - 8  of 14
  1  2  Következő