|
![]() |
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | PSAA |
Reactivity | A. thaliana |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Western blot | Yes |
Immunogen | N-terminal part of recombinant PsaA protein from Chlamydomonas reinhardtii P12154 |
Tested applications | Immuno-Gold |