Szeretne újra bejelentkezni?
Anti-NAPA Mouse monoclonal antibody unconjugated [clone: 4E204]
Gyártó: US Biological
Type: Primary
Antigen: NAPA (N-ethylmaleimide-sensitive factor attachment protein, alpha)
Clonality: Monoclonal
Clone: 4E+204
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Chicken, Dog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
RENDELÉS
Többet mutat / Kevesebbet mutat
100 µg 100 µG USBIS1014-75-100
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
Antigen symbol NAPA
Antigen name N-ethylmaleimide-sensitive factor attachment Protein, alpha
Reactivity Human, Mouse, Hamster, Pig, Bovine, Rabbit, Monkey, Dog, Sheep, Rat, Chicken
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 4E204
Host Mouse
Isotype IgG1 kappa
Western blot Yes
ImmunoPrecipitation Yes
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség