Szeretne újra bejelentkezni?
Anti-SNAI2 Mouse monoclonal antibody unconjugated [clone: 9H276]
Gyártó: US Biological
Recommended dilution: Western Blot: 1-2μg/ml
Type: Primary
Antigen: SNAI2 (snail family zinc finger 2)
Clonality: Monoclonal
Clone: 9H276
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
RENDELÉS
Többet mutat / Kevesebbet mutat
100 µg 100 µG USBIS1014-67M2
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
Antigen symbol SNAI2
Antigen name snail family zinc finger 2
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 9H276
Host Mouse
Isotype IgG1 kappa
Western blot Yes
Amino acid number 97 to 169
Purification purified by Protein G chromatography
Form liquid
Storage buffer 0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség