|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | SNAI |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Purification | purified by immunoaffinity chromatography |
Form | liquid |
Storage buffer | PBS, 0,09% sodium azide. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |