|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | SMURF2 |
Antigen name | SMAD specific E3 ubiquitin Protein ligase 2 |
Reactivity | Mouse, Human |
Conjugation | APC (Allophycocyanin) |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Epitope | N-terminal |
Purification | purified by Protein G chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, contains ~0,09% sodium azide |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |
Tested applications | Fluoresence ELISA |