|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | SMO |
Antigen name | smoothened, frizzled class receptor |
Reactivity | Bovine, Human, Dog |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Goat |
Western blot | Yes |
ELISA | Yes |
Koncentráció | ~0,5 mg/ml |
Purification | purified by immunoaffinity chromatography |
Szekvencia | C-NLMDTELMDADSD |
Form | liquid |
Storage buffer | Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |