|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | SMTN |
Antigen name | smoothelin |
Reactivity | Dog, Human, Cat, Monkey, Pig, Chicken |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody type | Primary |
Clone | 5F294 |
Host | Mouse |
Isotype | IgG1 |
Western blot | Yes |
ImmunoFluorescence | Yes |
Form | liquid |
Storage buffer | 0,09% sodium azide. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |