Szeretne újra bejelentkezni?
Anti-PTPRG Mouse monoclonal antibody unconjugated [clone: 8H210]
Gyártó: US Biological
Recommended dilution: Western Blot: 1 μg/ml
Type: Primary
Antigen: PTPRG (protein tyrosine phosphatase, receptor type, G)
Clonality: monoclonal
Clone: 8H210
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
  Remember me on this device
RENDELÉS
Többet mutat / Kevesebbet mutat
100 µg 100 µG USBIP9109-40H2
Vonalkód generálása Barcode Label Format: 
PDF katalógus oldal generálása
SPECIFIKÁCIÓK
Antigen symbol PTPRG
Antigen name Protein tyrosine phosphatase receptor type G
Reactivity Mouse, Human, Rat
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 8H210
Host Mouse
Isotype IgG2b
Western blot Yes
Amino acid number 111 to 541
Purification purified by Protein G chromatography
Form lyophilized powder
Storage buffer PBSwith 5% trehalose.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség