Szeretne újra bejelentkezni?
Anti-Protein Tyrosine Phosphatase, Non-receptor Type Substrate 1 Chicken polyclonal antibody unconjugated
Gyártó: US Biological
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Protein Tyrosine Phosphatase, Non-receptor Type Substrate 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
RENDELÉS
Többet mutat / Kevesebbet mutat
50 µg 50 µG USBIP9109-33
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
Antigen name Protein Tyrosine Phosphatase, Non-receptor Type Substrate 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Chicken
Isotype IgY
Western blot Yes
ELISA Yes
Amino acid number 252 to 360
Purification purified by immunoaffinity chromatography
Form liquid
Storage buffer PBS, pH 7,2, 0,02% sodium azide.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség