|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | PTPN13 |
Antigen name | Protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associated phosphatase) |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
ELISA | Yes |
Amino acid number | 2451 to 2466 |
Koncentráció | ~1 mg/ml |
Purification | purified by Protein G chromatography |
Form | liquid |
Storage buffer | PBS, 0,1% sodium azide. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |