Szeretne újra bejelentkezni?
Anti-PTPN9 Rabbit polyclonal antibody AP (Alkaline Phosphatase)
Gyártó: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:10-1:50
Do not freeze!
Type: Primary
Antigen: PTPN9 (protein tyrosine phosphatase, non-receptor type 9)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
RENDELÉS
Többet mutat / Kevesebbet mutat
200 µl 200 µl USBIP9109-28V-AP
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
Antigen symbol PTPN9
Antigen name Protein tyrosine phosphatase, non-receptor type 9
Reactivity Human
Conjugation AP (Alkaline Phosphatase)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitope C-terminal
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség