|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | ELP1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Amino acid number | 148 to 161 |
Koncentráció | ~1 mg/ml |
Purification | purified by immunoaffinity chromatography |
Form | liquid |
Storage buffer | PBS, 0,02% sodium azide. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |