|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | Elongation Factor RNA Polymerase II, 2 |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody type | Primary |
Clone | 8C129 |
Host | Mouse |
Isotype | IgG1 |
Western blot | Yes |
Koncentráció | ~0,1 mg/ml |
Purification | purified by Protein G chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,4, 1% BSA, 0,05% sodium azide. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |