|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | ELMO2 |
Antigen name | engulfment and cell motility 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Goat |
Western blot | Yes |
Epitope | C-terminal |
Koncentráció | ~0,5 mg/ml |
Purification | purified by immunoaffinity chromatography |
Szekvencia | C-PKEPSSYDFVYHYG |
Form | liquid |
Storage buffer | Tris saline buffer, pH 7,3, 0,5% BSA, 0,02% sodium azide. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |