|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | His |
Antigen name | His-tag |
Conjugation | FITC (Fluorescein Isothiocyanate) |
Clonality | Polyclonal |
Antibody type | Tag |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Purification | purified by immunoaffinity chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, contains ~0,09% sodium azide |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |
Tested applications | Fluoresence ELISA |