Szeretne újra bejelentkezni?
Anti-HIRIP3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Gyártó: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HIRIP3 (HIRA interacting protein 3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
RENDELÉS
Többet mutat / Kevesebbet mutat
200 µl 200 µl USBI036574-AP
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
Antigen symbol HIRIP3
Antigen name HIRA interacting Protein 3
Reactivity Human
Conjugation AP (Alkaline Phosphatase)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 286 to 316
Epitope internal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség