|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | HIPK4 |
Antigen name | homeodomain interacting Protein kinase 4 |
Reactivity | Mouse |
Conjugation | APC (Allophycocyanin) |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Amino acid number | 532 to 562 |
Epitope | C-terminal |
Purification | purified by Protein A chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, contains ~0,09% sodium azide |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |
Tested applications | Fluoresence ELISA |