Szeretne újra bejelentkezni?
Anti-HIPK3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Gyártó: US Biological
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: HIPK3 (homeodomain interacting protein kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
RENDELÉS
Többet mutat / Kevesebbet mutat
200 µl 200 µl USBI036568-AP
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
Antigen symbol HIPK3
Antigen name homeodomain interacting Protein kinase 3
Reactivity Human
Conjugation AP (Alkaline Phosphatase)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 1134 to 1163
Epitope C-terminal
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség