|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | HIPK3 |
Antigen name | homeodomain interacting Protein kinase 3 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Amino acid number | 1134 to 1163 |
Epitope | C-terminal |
Purification | purified by Protein G chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, 0,09% sodium azide. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |