Szeretne újra bejelentkezni?
Anti-HIP1R Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Gyártó: US Biological
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: HIP1R (huntingtin interacting protein 1 related)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
RENDELÉS
Többet mutat / Kevesebbet mutat
200 µl 200 µl USBI036562-PE
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
Antigen symbol HIP1R
Antigen name huntingtin interacting Protein 1 related
Reactivity Human
Conjugation PE (Phycoerythrin)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Flow cytometry Yes
Amino acid number 105 to 133
Epitope N-terminal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2
Tárolási hőmérséklet Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség