|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | HIP1R |
Antigen name | huntingtin interacting Protein 1 related |
Reactivity | Human |
Conjugation | AP (Alkaline Phosphatase) |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Flow cytometry | Yes |
Amino acid number | 105 to 133 |
Epitope | N-terminal |
Purification | purified by Protein A chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, contains ~0,09% sodium azide |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |