Szeretne újra bejelentkezni?
Anti-HINT3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Gyártó: US Biological
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HINT3 (histidine triad nucleotide binding protein 3)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
  Remember me on this device
RENDELÉS
Többet mutat / Kevesebbet mutat
200 µl 200 µl USBI036560-FITC
Vonalkód generálása Barcode Label Format: 
PDF katalógus oldal generálása
SPECIFIKÁCIÓK
Antigen symbol HINT3
Antigen name histidine triad nucleotide binding Protein 3
Reactivity Human
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 18 to 47
Epitope N-terminal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Tárolási hőmérséklet Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség