|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | LRRK2 |
Antigen name | leucine-rich repeat kinase 2 |
Reactivity | Human, Mouse |
Conjugation | Biotin |
Clonality | Monoclonal |
Antibody type | Primary |
Clone | 133AT1218 |
Host | Mouse |
Isotype | IgG1 |
Western blot | Yes |
ELISA | Yes |
Purification | purified by Protein G chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2 |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |