Szeretne újra bejelentkezni?
Anti-OR5B12 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 609CT8.2.4]
Gyártó: US Biological
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250
Type: Primary
Antigen: OR5B12 (olfactory receptor, family 5, subfamily B, member 12)
Clonality: Monoclonal
Clone: 609CT8.2.4
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human
  Remember me on this device
RENDELÉS
Többet mutat / Kevesebbet mutat
100 µl 100 µl USBI031174-APC
Vonalkód generálása Barcode Label Format: 
PDF katalógus oldal generálása
SPECIFIKÁCIÓK
Antigen symbol OR5B12
Antigen name olfactory receptor, family 5, subfamily B, member 12
Reactivity Human
Conjugation APC (Allophycocyanin)
Clonality Monoclonal
Antibody type Primary
Clone 609CT8.2.4
Host Mouse
Isotype IgG2b
Western blot Yes
ELISA Yes
Epitope C-terminal
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség