Szeretne újra bejelentkezni?
Anti-DUSP1 Rabbit Polyclonal Antibody
Gyártó: OriGene
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000
Type: Primary
Antigen: DUSP1 (dual specificity phosphatase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
RENDELÉS
Többet mutat / Kevesebbet mutat
100 µl 100 µl ORIGTA313772
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
Antigen symbol DUSP1
Antigen name Dual specificity phosphatase 1
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Modification phospho-Ser359
Cross adsorption none
Purification purified by affinity chromatography
Form Liquid
Storage buffer Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség