Szeretne újra bejelentkezni?
Anti-DPYSL2 Rabbit Polyclonal Antibody
Gyártó: OriGene
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000
Type: Primary
Antigen: DPYSL2 (dihydropyrimidinase-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
  Remember me on this device
RENDELÉS
Többet mutat / Kevesebbet mutat
100 µl 100 µl ORIGTA313765
Vonalkód generálása Barcode Label Format: 
PDF katalógus oldal generálása
SPECIFIKÁCIÓK
Antigen symbol DPYSL2
Antigen name Dihydropyrimidinase-like 2
Reactivity Human, Rat, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Cross adsorption none
Purification purified by affinity chromatography
Form Liquid
Storage buffer Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség