|
|
RENDELÉS
SPECIFIKÁCIÓK
Antigen symbol | CREBBP |
Antigen name | CREB binding protein |
Reactivity | Human, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
Cross adsorption | none |
Purification | purified by affinity chromatography |
Form | Liquid |
Storage buffer | Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol. |
Tárolási hőmérséklet | Cool |
Shipping temperature | Ice |