  • Szűrés gyártók szerint
    Rendezés:
  • Keresés szűkítése
Genomikai DNS tisztító készletek
https://hu.vwr.com/stibo/smallweb/std.lang.all/52/92/18945292.jpg
For high-quality genomic DNA purification starting materials will need special lysis conditions and might contain extremely low concentrations of DNA, or high levels of inhibitors. VWR’s portfolio offers just the right solution for almost any sample type, including cells or tissues from various animals or plants, biological liquids, forensic specimens, stool or soil samples. Purified DNA gathered from these kits is suitable for direct use in all common molecular biology applications, such as PCR, restriction digestion, cloning, hybridization, and sequencing.
SearchPresentationType-HORIZONTAL
SearchResultCount:"102"
Rendezés:
Gyártó:  VWR Peqlab
Leírás:   For isolation of RNA, DNA and protein from one sample. Contains phenol and guanidine isothiocyanate.
Gyártó:  VWR Chemicals
Leírás:   The Yeast Genomic DNA Purification Kit yields high molecular weight DNA from yeast using a non-enzymatic lysis buffer and no glass beads. The protocol...
Katalógus szám: (1B1558-KIT)
Gyártó:  VWR Chemicals
Leírás:   Fluorescent DNA Quantitation Kit uses fluorescent bisBenzimide H33258 dye to bind to AT sequences of the minor groove of double-stranded DNA. In the a...
UOM:  1 * 1 KIT
Gyártó:  VWR Peqlab
Leírás:   Plant DNA Mini Kit for fast and efficient extraction of high quality DNA from nucleus, plastids and mitochondria of a wide range of plant species and ...
Gyártó:  VWR Peqlab
Leírás:   Blood DNA Mini Kit Plus offers a safe, quick and efficient method for extracting pure and high quality DNA from fresh, frozen and anticoagulated treat...
Gyártó:  VWR Peqlab
Leírás:   Tissue DNA Mini kits for the quick and reliable isolation of high quality DNA from a wide range of animal and human cells and tissue. An optimised Mic...
Katalógus szám: (K368-50RXN)
Gyártó:  VWR Chemicals
Leírás:   Cyclo-Prep™ Genomic DNA Purification Kit efficiently isolates genomic DNA for PCR amplification, RFLP analysis, Southern blotting, DNA fiingerprinting...
UOM:  1 * 50 DB
Gyártó:  VWR Peqlab
Leírás:   Bacterial DNA Mini Kit for the extraction of high quality DNA from gram positive and gram negative bacteria. For up to 1×109 cells per prep...
1 - 8  of 102
  1  2  3  4  5  6  7  8  9  10  11  12  13  Következő