Szeretne újra bejelentkezni?
Baskets, mobile injectors, mobile units, covers and injector nozzles for laboratory glassware washing machines
Gyártó: Miele
RENDELÉS
Többet mutat / Kevesebbet mutat
Basket
Leírás M.egys Szél. × mélys. × mag. Dla VWR katalógus szám Mennyiség
O 175, felső injektoros kosár szűknyakú üvegeszközökhöz 1 531×475×250 mm PG 8535, PG 8536 141-0345
O 184, felső injektoros kosár különféle laboratóriumi üvegeszközökhöz 1 531×475×468 mm PG 8535/36 141-0346
O 187, felső injektoros kosár szűknyakú üvegeszközökhöz 1 531×475×250 mm PG 8535/36 141-0392
U 188/1, felső kosár/illeszték több betéthez 1 531×475×215 mm PG 8535/36 141-0562
U 175/1, alsó injektoros kosár szűknyakú üvegeszközökhöz 1 531×475×250 mm PG 8536 141-0515
U 184/1, alsó injektoros kosár különféle laboratóriumi üvegeszközökhöz 1 531×515×250 mm PG 8536 141-0516
A 150 lower basket for misc. inserts 1 - PG 8583, PG 8583 CD, PG 8593 141-1258
O 190/2 upper basket with open front, empty, for use with various inserts, integrated spray arm 1 531×475×265 mm PG 8535/36 141-0341
E 380, teljes kosár szűknyakú üvegeszközökhöz 1 535×517×502 mm PG 8535/36 141-0366
U 874/1, alsó kosár/illeszték több betéthez 1 531×515×50 mm PG 8535/36 141-0517
E 329, teljes injektoros kosár szűknyakú üvegeszközökhöz 1 535×517×502 mm PG 8535/36 141-0357
E 350, teljes injektoros kosár szűknyakú üvegeszközökhöz 1 535×517×502 mm PG 8535/36 141-0361
A 100 upper basket with 2 docking pipes for injector modules or inserts 1 525×528×141 mm PG 8583, PG 8583 CD, PG 8593 141-1255
A 101 upper basket for various inserts, height adjustable, with built-in spray arm 1 528×527×206 mm PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8592, PG 8593 141-1256
A 102 upper basket for various inserts, height adjustable, with built-in spray arm 1 528×527×206 mm PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593 141-1257
A 103 upper basket for various inserts, with built-in spray arm 1 528×528×123 mm PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8592, PG 8593 MIEL9862370
A 151 lower basket for misc. inserts 1 529×522×88 mm PG 8581, PG 8582, PG 8582CD, PG 8583, PG 8583CD, PG 8591, PG 8592, PG 8593 141-1259
O 191/2 upper basket with open front, for mesh trays, integrated spray arm 1 531×475×180 mm PG 8535, PG 8536 141-0563
O 800/1 upper basket, for various inserts, integrated spray arm 1 381×475×270 mm G 7831 141-0397
O 801/2 upper basket, central axis with grid for hollow instruments and 10 nozzles, for various inserts, integrated spray arm 1 381×475×267 mm G 8731 141-0407
Mobile injector
Leírás M.egys Szél. × mélys. × mag. Dla VWR katalógus szám Mennyiség
E 743 injector module for narrow necked glassware 100 - 500 ml, 36 injector nozzles (E 351), 4×160 mm 1 492×496×190 mm G 7825 141-0420
E 744 injector module for narrow necked glassware 500 - 1000 ml, 16 injector nozzles (E 352), 6×220 mm 1 492×496×250 mm G 7825 141-0421
E 752 injector module for narrow necked glassware 100 - 1000 ml, 25 injector nozzles (12× E 351, 4×160 mm and 13× E 352, 6×220 mm) 1 492×496×260 mm G 7825 141-0426
E 755 injector module for narrow necked glassware 25 - 100 ml, 36 injector nozzles (E 470), 2,5×90 mm 1 492×496×130 mm G 7825 141-0433
E 757 mobile unit with drying connection, for 1 - 6 large items (ex. carboys): 6 injector nozzles with plastic star bases, height adjustable holding frame with 6 short and 4 long struts for various glass diameters 1 530×600×346 mm G 7825 141-0440
E 943/2 injector module for narrow neck items 100 - 500 ml, 32 nozzles (E 351) 4×160 mm with clips (E 353) 1 558×352×190 mm PG 8527 141-0574
E 944/2 injector module for narrow-necked glassware 500 - 1000 ml, 15 nozzles (E 352) 6×220 mm with clips (E 354) 1 558×352×250 mm PG 8527 141-0575
E 957 mobile unit with drying connection, for 1 - 12 large volume glassware: 12 injector nozzles (Ø 10 mm), height-adjustable holding frame with 8 short and 6 long supports and supports can be adjusted to accommodate various glass diameters
Magnetic strip for automatic mobile unit recognition
1 640×790×353 mm PG 8527 141-0441
Injector for butyrometer SD-B, for mobile injector unit E 331 1 — mm G 7804, G 7883, G 7893, G 7883 CD, G 7835 CD, G 7836 CD 141-0344
A 300 injector module with 8 injector nozzles (E 352), for laboratory glassware (Erlenmeyer flasks, flasks, bottles etc.,) 1 200×471×241 mm PG 8583, PG 8583 CD, PG 8593 141-1261 Unavailable
A 301/2 injector module with 3×6 injector nozzles (ID 160), for laboratory glassware (Erlenmeyer flasks, flasks, bottles etc.,) 1 220×479×181 mm PG 8583, PG 8583 CD, PG 8593 141-1265 Unavailable
A 302/1 injector module with 4×8 injector nozzles (E 351), for laboratory glassware (Erlenmeyer flasks, flasks, bottles etc.,) 1 235×479×181 mm PG 8583, PG 8583 CD, PG 8593 141-1268 Unavailable
A 302 injector module with 4×8 injector nozzles (ID 110), for laboratory glassware (Erlenmeyer flasks, flasks, bottles etc.,) 1 215×479×132 mm PG 8583, PG 8583 CD, PG 8593 141-1267 Unavailable
A 303 injector module for 98 pipettes 1 222×471×185 mm PG 8583, PG 8583 CD, PG 8593 141-1269
A 304 injector module for 98 phials 1 222×471×130 mm PG 8583, PG 8583 CD, PG 8593 141-1270
A 306 module for measuring cylinders (4×2 l) 1 418×471×235 mm PG 8583, PG 8583 CD, PG 8593 141-1271
E 742 module frame for miscellaneous glassware, for use in mobile unit E 741/2, integrated spray arm 1 492×496×112 mm G 7825 141-0472
E 746 injector module for 23 pipettes (10 pipettes up to 560 mm and 13 pipettes up to 490 mm), allows a second cleaning level 1 492×496×330 mm G 7825 141-0422
E 747 injector module, for cleaning centrifuge tubes, vials, test tubes, or other small items, with forced-air drying through injectors, 104 injector nozzles, 2,5×110 mm 1 492×496×168 mm G 7825 141-0416
E 900-4/2 mobile unit with drying unit (empty), for inserts on 4 levels, with 3 built-in spray arms
Magnetic strip for automatic mobile unit recognition
1 640×790×557 mm PG 8527, PG 8528 141-0586
E 900-5/2 mobile unit with drying unit (empty), for inserts on 5 levels, 4 built-in spray arms
Magnetic strip for automatic
mobile unit recognition
1 640×790×605 mm PG 8527, PG 8528 141-0587
E 935/2 load carrier with drying connection (empty), for inserts on 3 levels, with 2 built-in spray arms
Magnetic strip for automatic mobile unit recognition
1 640×790×524 mm PG 8527, PG 8528 141-0588
E 940 mobile unit with drying connection, narrow-necked glassware on 2 levels (the mobile injector unit can be used without upper level)
115 nozzles with clips: 35 nozzles (E 352) 6×220 mm with clips (E 354) for the lower level, 80 nozzles (E 351) 4×160 mm with clips (E 353) for the upper level
Magnetic strip for automatic mobile unit recognition
1 640×790×565 mm PG 8527 141-0396
E 942/3 injector module for 116 pipettes up to 580 mm, with holder frame, compartment size 16×16 mm 1 558×352×279 mm PG 8527 141-0573
E 945/2 injector module for misc. Inserts 1 558×352×55 mm PG 8527 141-0576
E 947/2 injector module with 88 injector nozzles for centrifuge tubes, phials and fraction
sampler tubes
88 nozzles 2,5 ×110 mm
1 558×352×170 mm G 8527 141-0577
E 950/1 mobile unit with drying connection, for narrow-necked glassware on 3 levels, with 232 injector nozzles
Levels 1 and 3: 80 nozzles ID 90 each
Level 2: 72 nozzles ID 90 each
Max. loading height 148 mm on all 3 levels
1 640×790×572 mm PG 8527 141-0526
E 975/2 injector module for inserts on 2 levels, with integrated spray arm
Magnetic strip for automatic mobile unit recognition
1 640×790×427 mm PG 8527, PG 8528 141-0589
Modular injector insert E 745/1, for 104 pipettes, each section is 16×16 mm, with forced-air drying through injectors
Frame supports wide mouth glassware
1 492×496×288 mm G 7825 and G 7826 141-0508
Mobile units
Leírás M.egys Szél. × mélys. × mag. Dla VWR katalógus szám Mennyiség
E 404/1, teljes injektoros kosár 38 pipettához 1 535×517×502 mm PG 8535/36 141-0527
E 405/1 mobile injector unit with drying connection, for 38 pipettes in 3 rows
row 1: 10 pipettes 100 ml (length up to 550 mm), spacing 20 mm
row 2: 14 pipettes 25 ml, spacing 26 mm
row 3: 14 pipettes 10 ml, spacing 26 mm
With holder for ML/2 magnetic strip for automatic mobile unit recognition and integrated spray arm
1 535×517×502 mm PG 8535/36 141-0336
E 406, teljes injektoros kosár 116 pipettához 1 535×517×502 mm PG 8535/36 141-0368
E 408, teljes injektoros kosár 96 pipettához 1 535×517×502 mm PG 8535/36 141-0369
E 355, fél injektoros kosár szűknyakú üvegeszközökhöz 1 535×517×502 mm PG 8535/36 141-1000
E 414, teljes injektoros kosár szűknyakú üvegeszközökhöz 1 535×517×502 mm PG 8535/36 141-0370
E 331, mobil injektoros kosár butirométerekhez 1 — mm PG 8535/36 141-0358
E 340, fél injektoros kosár szűknyakú üvegeszközökhöz 1 — mm PG 8535/36 141-0360
A 200 mobile unit for 38 pipettes 1 529×546×397 mm PG 8583, PG 8593 141-1260
E 385 injector unit with drying unit connection, for narrow-necked glassware, with 16 injector nozzles, with holder for ML/2 magnetic strip for automatic mobile unit recognition
Supplied as standard with: 7×E 351 nozzles (Ø 4,0×160 mm), 7×E 353 clips for injector nozzles (Ø 4,0×160 mm), 9×E 352 nozzles (Ø 6,0×220 mm), 9×E 354 clips for injector nozzles (Ø 6,0×220 mm), 1× irrigation nozzle for powder dispenser
1 535×517×502 mm PG 8535/36 141-0367
E 741/1 injector unit, empty for modules in 1-4 levels, up to 3 modules can be accommodated depending on size of items (water and air inlets connected to the basket direct, to the modules via adapter)
Loading dimensions: Level 1 to top edge: 605 mm / Level 2 to top edge: 405 mm / Level 3 to top edge: 267 mm / Level 4 to top edge: 197 mm
1 530×600×680 mm G 7825 141-0490
E 775/1 mobile unit (empty), for inserts on 2 levels, with built-in spray arm and magnetic strip for automatic mobile unit recognition (excl. ML magnets) 1 530×600×400 mm G 7825, G 7826 141-0585
E 941 injector module with 2 levels for modular inserts (up to 2 modules can be accommodated on each level depending on size of items), water and drying air enters via a direct docking system and adapters 1 619×790×421 mm PG 8527 141-0398
Covers
Leírás M.egys Szél. × mélys. × mag. Dla VWR katalógus szám Mennyiség
A 2, cover plastic netted with metal frame ,for 1/2 inserts 1 216×456×0 mm PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0389
A 13 stainless steel lid for quarter inserts E 103/1, E 104/1, E 105/1 and E 139/1 1 200×320×20 mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0329
A 3 cover plastic netted with metal frame ,for 1/4 inserts 1 206×206×0 mm G 7825, PG 8527,PG 8535/36, PG 8583, PG 8583 CD 141-0390
A 5 cover plastic netted with stainless steel frame, for baskets O 184, U 184 1 280×280×8 mm PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0425
A 14 lid 1/4 for AK 12 insert, in stainless steel
7×7 mm perforations, 3 mm ridge
1 210×210×20 mm PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0394
A 19 perforated metal lid, fits to insert E 969 1 251×351×18 mm PG 8527 141-0438
Stainless steel lid, DE-CS6-78 1 900×600×30 mm G 7883 141-0437
Stainless steel lid, DE-CS7-78 1 900×600×30 mm G 7883 CD 141-0291
Stainless steel lid, DE-CS7-85 1 - mm PG 8583 CD 141-1403
Injector nozzles
Leírás M.egys Szél. × mélys. × mag. Dla VWR katalógus szám Mennyiség
ID 180 injector nozzle with plastic support
Ø: 4×180 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0375
ID 220 injector nozzle with plastic support
Ø: 6×220 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0377
E 470 injector nozzle with holders for injector unit
Ø×H: 2,5×90 mm, screw connection
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0432
ID 240 injector nozzle with plastic support
Ø: 6×240 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0378
ID 200 injector nozzle with plastic support
Ø: 6×200 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0376
ID 110 injector nozzle with plastic support
Ø: 2,5×110 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0372
ID 160 injector nozzle with plastic support
Ø: 4×160 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0374
ID 90 injector nozzle with plastic support
Ø: 2,5×90 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0371
E 354 clip for injector nozzle E352, height adjustable
Ø×H: 6×220 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0365
E 353 clip for injector nozzle E351, height adjustable
Ø×H: 4×160 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0364
E 352 injector nozzle for mobile injector unit; in conjunction with E 354
Ø×H: 6×220 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0363
E 351 injector nozzle for mobile injector unit; in conjunction with E 353
Ø×H: 4×160 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0362
E 336 injector sleeve, for pipettes (max. length 445 mm) in injector units, plastic with screw fitting
Ø 11 mm, length 121 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8592, PG 8593 141-0359
ID 140 injector nozzle with plastic support
Ø: 4×140 mm
1 — mm G 7825, PG 8527, PG 8535/36, PG 8583, PG 8583 CD, PG 8593 141-0373
unavailable Ez a termék már nem elérhető. Valószínű, hogy vannak elérhető alternatívák ha rákeres az alább megadott VWR katalógus számra. Ha további segítségre lenne szüksége kérjük hívja vevőszolgálatunkat az (52) 521-130 telefonszámon.
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
SPECIFIKÁCIÓK
7835CD
7836CD
7883CD
Magasság, mm
G 7893
Típus
Alsó kosárhoz
Felső kosárhoz
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség