Szeretne újra bejelentkezni?
Accessories
Gyártó: Binder
RENDELÉS
Többet mutat / Kevesebbet mutat
Leírás M.egys Dla VWR katalógus szám Mennyiség
2 Doors with windows and interior lighting, 470×290 mm 1 FP range BIND8012-0062
Access port on left hand side with 30 mm Ø silicone plug 1 All models 390-0582
Access port with 10 mm Ø silicone plug, at rear 1 BD,BF,ED,FD,FED,FP,M, FDL incubators BIND8012-0390
Access port with 10 mm Ø silicone plug, left hand side 1 BD,BF,ED,FD,FED,FP,M, FDL incubators BIND8012-0388
Access port with 10 mm Ø silicone plug, on top 1 BD,BF,ED,FD,FED,FP,M incubators BIND8012-0386
Access port with 10 mm Ø silicone plug, right hand side 1 BD,BF,ED,FD,FED,FP,M, FDL incubators 466-0170
Access port with 100 mm Ø silicone plug, left hand side 1 ED,FD,FED,FP,M incubators BIND8012-0722
Access port with 100 mm Ø silicone plug, on top 1 BD, BF, KB, KT incubators BIND8012-0053
Access port with 100 mm Ø silicone plug, on top 1 ED,FD,FED,FP incubators BIND8012-0547
Access port with 100 mm Ø silicone plug, right hand side 1 ED,FD,FED,FP,M incubators BIND8012-0721
Access port with 29 mm Ø silicone plug, left hand side 1 BD, BF, KB, KT incubators 466-0485
Access port with 29 mm Ø silicone plug, on top 1 BD, BF, KB, KT incubators 466-0156
Access port with 29 mm Ø silicone plug, right hand side 1 BD, BF, KB and KT incubators 466-0157
Access port with 30 mm Ø silicone plug, at rear 1 BD incubators BIND8012-0397
Fan, stronger, to increase air change rate 1 FED 53, FP 53 models BIND8012-0197
Access port with 30 mm Ø silicone plug, at rear 1 ED,FD,FED,FP,M incubators BIND8012-0540
Fan, stronger, to increase air change rate 1 FED 240, FP 240 models BIND8012-0199
Access port with 30 mm Ø silicone plug, on top 1 ED,FD,FED,FP,M incubators BIND8012-0539
Access port with 50 mm Ø silicone plug, at rear 1 BD 720 BIND8012-0399
Access port with 50 mm Ø silicone plug, at rear 1 ED,FD,FED,FP,M incubators BIND8012-0546
Access port with 50 mm Ø silicone plug, left hand side 1 BD, BF, KT incubators BIND8012-0051
Access port with 50 mm Ø silicone plug, left hand side 1 ED,FD,FED,FP,M incubators BIND8012-0542
Access port with 50 mm Ø silicone plug, on top 1 BD, BF, KB, KT incubators BIND8012-0049
Access port with 50 mm Ø silicone plug, on top 1 ED,FD,FED,FP,M incubators BIND8012-0545
Access port with 50 mm Ø silicone plug, right hand side 1 BD, BF, KT incubators 466-0158
Inner chamber, reinforced, stainless steel, with 2 racks, max total load of 250 kg, max load per rack 70 kg 1 FED incubators BIND8012-0338
Access port with 50 mm Ø silicone plug, right hand side 1 ED,FD,FED,FP,M incubators BIND8012-0541
Access port with silicone plug 30 mm, right side towards top 1 ED,FD,FED,FP,M,FDL incubators BIND8012-0536
Base on castors 1 BD,BF,M incubators BIND9051-0033
Over temperature alarm, temperature limit can be set at the independent, adjustable temperature safety device for 230V 1 BD,BF,ED,FD,FED,FP incubators BIND8012-0469
Base on castors, with lockable adjustable feet, W×D×H : 700×744×129 1 C 150 series 390-6072
Door gasket, FKM 1 ED,FD,FED,FP,M incubators BIND8012-0495
Door gasket, FKM (viton) 1 FED 53, FP 53, M 53, ED 53, FD 53 models BIND8012-0494
Door with window 180×180 mm, interior lighting (15W) 400V 3N ~ 60Hz 1 ED023 BIND8012-0378
Door with window 320×260 mm and interior lighting (15W) 1 FP 115 model 466-0159
Door with window 320×260 mm, interior lighting (15W) 230V 1N ~ 50/60Hz 1 FD,FED incubators BIND8012-0215
Door with window 350×240 mm, interior lighting (30W) 230V 1N ~ 50/60Hz 1 FED incubators BIND8012-0216
Door with window 470×290 mm, interior lighting (30W) 400V 3N ~ 60Hz 1 ED400 BIND8012-0269
Door with window 470×290 mm, interior lighting (30W) 400V 3N ~ 60Hz 1 ED720 BIND8012-0270
USB cable/RS422 interface converter 1 All Binder models 390-0717
Heavy duty rack, stainless steel, maximum load per rack 70 kg 1 BD/ED 720 466-0512
HEPA filter, class EU 14H 14 1 FP range temperature test chambers 466-0161
Interior socket, waterproof, coverable with associated LEMO connector (nominal voltage, max. 500 W, max. 90 °C, protection class IP67) 1 KB, KBW, KBF models BIND8012-0640
Perforated shelf, stainless steel 1 BD/ED 720 466-0506
Rack lock 1 KIT FP range temperature test chambers 391-0072
Rack, reinforced, stainless steel, with 1 set of 4 fasteners, max load of 70 kg 1 FED 400, FP 400, M 400 incubators BIND8012-0346
Rack, chrome-plated 1 BD/ED 720 466-0510
Rack, stainless steel 1 BD/ED 720 466-0508
Stable table on wheels with castors and locking brakes 1 BD, ED, FD models BIND9051-0018
Factory calibration certificate, measurement taken in centre of chamber or at a specified testing temperature 1 C,CB,B,BF,BD,KT,KB,KBW,E,ED,FED,FP,M,FDL,MK, MKT incubators 466-0155
Independent temperature safety device, Class 3.1, with visual alarm 1 ED,FD,FED,FP incubators BIND8012-0032
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség