Szeretne újra bejelentkezni?
Capillary columns, VF-200ms
Gyártó: Agilent Technologies
VF-200ms is designed with a unique selectivity for compounds rich in dipole-dipole interactions, resulting from the electrophilic nature of the trifluoropropyl stationary phase.
  • Superior deactivation delivers symmetrical peaks to improve data accuracy
  • Ultra-low background noise for trace analysis maximizes sensitivity
  • Ideal for sensitive and selective detector systems for enhanced productivity
  • Suited for electron rich, high dipole moment compounds like ketones, aldehydes, nitro- or chloro-containing compounds, PAHs, unsaturated compounds, silanes and CFCs
  • Accurate peak identification and reliable analysis
The VF-200ms trifluoropropyl phase has very high temperature stability and can be used routinely up to 350 °C.
RENDELÉS
Többet mutat / Kevesebbet mutat
Kolonna formája Fázis Rétegvastagság Belső átmérő × hossz M.egys VWR katalógus szám Mennyiség
Capillary VF-200ms 0,15 µm 0,15 mm × 20 m 1 VARICP5891
Capillary VF-200ms 0,60 µm 0,15 mm × 20 m 1 VARICP5892
Capillary VF-200ms 0,10 µm 0,25 mm × 30 m 1 VARICP8857
Capillary VF-200ms 0,25 µm 0,25 mm × 30 m 1 VARICP8858
Capillary VF-200ms 0,50 µm 0,25 mm × 30 m 1 VARICP8859I5
Capillary VF-200ms 0,50 µm 0,25 mm × 30 m 1 VARICP8859
Capillary VF-200ms 1,00 µm 0,25 mm × 30 m 1 VARICP8860
Capillary VF-200ms 0,50 µm 0,32 mm × 30 m 1 VARICP8864
Capillary VF-200ms 1,00 µm 0,32 mm × 30 m 1 VARICP8865
Capillary VF-200ms 0,50 µm 0,53 mm × 30 m 1 VARICP8867
Capillary VF-200ms 1,00 µm 0,53 mm × 30 m 1 VARICP8868
Capillary VF-200ms 0,25 µm 0,25 mm × 60 m 1 VARICP8861
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség