Szeretne újra bejelentkezni?
Capillary columns, VF-5ms
Gyártó: Agilent Technologies
VF-5ms is a highly inert 5% phenyl-methyl column for increased sensitivity, accuracy and instrument uptime.
  • Excellent selectivity for aromatic compounds
  • Minimal column bleed improves sensitivity
  • Individual test certificates guarantee performance
  • Applicable for a wide range of semi-polar and even polar components, such as phenols
The columns have the lowest guaranteed bleed specification of 1 pA @ 325 °C (30 m, 0,25 mm, 0,25 µm).
RENDELÉS
Többet mutat / Kevesebbet mutat
Kolonna formája Fázis Rétegvastagság Belső átmérő × hossz M.egys VWR katalógus szám Mennyiség
Capillary VF-5ms 0,40 µm 0,10 mm × 10 m 1 VARICP8934
Capillary VF-5ms 0,15 µm 0,15 mm × 10 m 1 VARICP9034
Capillary VF-5ms 0,33 µm 0,20 mm × 12 m 1 VARICP8935
Capillary VF-5ms 0,10 µm 0,25 mm × 15 m 1 VARICP8938
Capillary VF-5ms 0,25 µm 0,25 mm × 15 m 1 VARICP8939
Capillary VF-5ms 1,00 µm 0,25 mm × 15 m 1 VARICP8940
Capillary VF-5ms 0,10 µm 0,32 mm × 15 m 1 VARICP8950
Capillary VF-5ms 0,25 µm 0,32 mm × 15 m 1 VARICP8951
Capillary VF-5ms 0,50 µm 0,25 mm × 15 m 1 VARICP8963
Capillary VF-5ms 0,50 µm 0,53 mm × 15 m 1 VARICP8971
Capillary VF-5ms 0,15 µm 0,15 mm × 15 m 1 VARICP9035
Capillary VF-5ms 0,15 µm 0,15 mm × 20 m 1 VARICP9036
Capillary VF-5ms 0,30 µm 0,15 mm × 20 m 1 VARICP9037
Capillary VF-5ms 0,60 µm 0,15 mm × 20 m 1 VARICP9038
Capillary VF-5ms 0,33 µm 0,20 mm × 25 m 1 VARICP8936
Capillary VF-5ms 0,25 µm 0,25 mm × 25 m 1 VARICP8941
Capillary VF-5ms 0,52 µm 0,32 mm × 25 m 1 VARICP8953
Capillary VF-5ms 0,10 µm 0,25 mm × 30 m 1 VARICP8943
Capillary VF-5ms 0,25 µm 0,25 mm × 30 m 1 VARICP8944I5
Capillary VF-5ms 0,25 µm 0,25 mm × 30 m 1 VARICP8944
Capillary VF-5ms 0,50 µm 0,25 mm × 30 m 1 VARICP8945
Capillary VF-5ms 1,00 µm 0,25 mm × 30 m 1 VARICP8946
Capillary VF-5ms 0,25 µm 0,32 mm × 30 m 1 VARICP8955
Capillary VF-5ms 0,50 µm 0,32 mm × 30 m 1 VARICP8956
Capillary VF-5ms 1,00 µm 0,32 mm × 30 m 1 VARICP8957
Capillary VF-5ms 0,50 µm 0,53 mm × 30 m 1 VARICP8974
Capillary VF-5ms 1,00 µm 0,53 mm × 30 m 1 VARICP8975
Capillary VF-5ms 1,50 µm 0,53 mm × 30 m 1 VARICP8976
Capillary VF-5ms 0,15 µm 0,15 mm × 40 m 1 VARICP9039
Capillary VF-5ms 0,33 µm 0,20 mm × 50 m 1 VARICP8937
Capillary VF-5ms 0,25 µm 0,25 mm × 50 m 1 VARICP8947
Capillary VF-5ms 0,25 µm 0,32 mm × 50 m 1 VARICP8958
Capillary VF-5ms 0,40 µm 0,32 mm × 50 m 1 VARICP8959
Capillary VF-5ms 0,10 µm 0,25 mm × 60 m 1 VARICP8948
Capillary VF-5ms 1,00 µm 0,25 mm × 60 m 1 VARICP8949
Capillary VF-5ms 0,25 µm 0,25 mm × 60 m 1 VARICP8960
Capillary VF-5ms 0,25 µm 0,32 mm × 60 m 1 VARICP8961
Capillary VF-5ms 1,00 µm 0,32 mm × 60 m 1 VARICP8962
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség