Szeretne újra bejelentkezni?
Capillary Columns, VF-1ms
Gyártó: Agilent Technologies
VF-1ms is a highly inert, non-polar, low bleed GC column providing increased sensitivity over a broad array of applications.
  • Lowest guaranteed bleed specification for trace analysis with MS
  • Wide range of applications ensures near universal applicability
  • Highly inert for accurate analysis, even at trace levels
100% dimethylpolysiloxane
Temperature limits: -60 to 325/350 °C
Similar phases: Rtx-1ms, Rxi-1ms, MDN-1, AT-1, ZB-1ms, Equity-1
RENDELÉS
Többet mutat / Kevesebbet mutat
Leírás Kolonna formája Fázis Rétegvastagság Belső átmérő × hossz M.egys VWR katalógus szám Mennyiség
on 7" cage Capillary VF-1ms 0,10 µm 0,10 mm × 10 m 1 568-0718
on 7" cage Capillary VF-1ms 0,40 µm 0,10 mm × 10 m 1 568-0719
on 7" cage Capillary VF-1ms 0,10 µm 0,10 mm × 20 m 1 568-0720
on 7" cage Capillary VF-1ms 0,40 µm 0,10 mm × 20 m 1 568-0721
on 7" cage Capillary VF-1ms 0,15 µm 0,15 mm × 10 m 1 568-0753
on 7" cage Capillary VF-1ms 0,15 µm 0,15 mm × 15 m 1 568-0697
on 7" cage Capillary VF-1ms 0,15 µm 0,15 mm × 20 m 1 568-0754
on 7" cage Capillary VF-1ms 0,60 µm 0,15 mm × 20 m 1 568-0755
on 7" cage Capillary VF-1ms 0,33 µm 0,20 mm × 12 m 1 568-0722
on 7" cage Capillary VF-1ms 0,33 µm 0,20 mm × 25 m 1 568-0723
on 7" cage Capillary VF-1ms 0,10 µm 0,25 mm × 15 m 1 568-0724
on 7" cage Capillary VF-1ms 0,25 µm 0,25 mm × 15 m 1 568-0725
on 7" cage Capillary VF-1ms 1,00 µm 0,25 mm × 15 m 1 568-0726
on 7" cage Capillary VF-1ms 0,25 µm 0,25 mm × 25 m 1 568-0728
on 7" cage Capillary VF-1ms 0,40 µm 0,25 mm × 25 m 1 568-0729
on 7" cage Capillary VF-1ms 0,10 µm 0,25 mm × 30 m 1 568-0730
on 7" cage Capillary VF-1ms 0,25 µm 0,25 mm × 30 m 1 568-0731
on 7" cage Capillary VF-1ms 1,00 µm 0,25 mm × 30 m 1 568-0733
on 7" cage Capillary VF-1ms 0,25 µm 0,25 mm × 50 m 1 568-0734
on 7" cage Capillary VF-1ms 0,40 µm 0,25 mm × 50 m 1 568-0735
on 7" cage Capillary VF-1ms 0,25 µm 0,25 mm × 60 m 1 568-0736
on 7" cage Capillary VF-1ms 1,00 µm 0,25 mm × 60 m 1 568-0737
on 7" cage Capillary VF-1ms 0,25 µm 0,32 mm × 15 m 1 568-0738
on 7" cage Capillary VF-1ms 0,25 µm 0,32 mm × 25 m 1 568-0739
on 7" cage Capillary VF-1ms 0,40 µm 0,32 mm × 25 m 1 568-0740
on 7" cage Capillary VF-1ms 0,10 µm 0,32 mm × 30 m 1 568-0741
on 7" cage Capillary VF-1ms 0,25 µm 0,32 mm × 30 m 1 568-0742
on 7" cage Capillary VF-1ms 0,50 µm 0,32 mm × 30 m 1 568-0743
on 7" cage Capillary VF-1ms 1,00 µm 0,32 mm × 30 m 1 568-0744
on 7" cage Capillary VF-1ms 0,40 µm 0,32 mm × 50 m 1 568-0745
on 7" cage Capillary VF-1ms 0,25 µm 0,32 mm × 60 m 1 568-0746
on 7" cage Capillary VF-1ms 1,00 µm 0,32 mm × 60 m 1 568-0747
on 7" cage Capillary VF-1ms 0,50 µm 0,53 mm × 15 m 1 568-0748
on 7" cage Capillary VF-1ms 1,50 µm 0,53 mm × 15 m 1 568-0749
on 7" cage Capillary VF-1ms 0,50 µm 0,53 mm × 30 m 1 568-0750
on 7" cage Capillary VF-1ms 1,00 µm 0,53 mm × 30 m 1 568-0751
on 7" cage Capillary VF-1ms 1,50 µm 0,53 mm × 30 m 1 568-0752
on 5" cage Capillary VF-1ms 1,00 µm 0,25 mm × 15 m 1 568-0727
on 5" cage Capillary VF-1ms 0,25 µm 0,25 mm × 30 m 1 568-0732
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség