Szeretne újra bejelentkezni?
Strong cation exchange HPLC columns, PL-SCX
Gyártó: Agilent Technologies
Macroporous PS/DVB matrix with a very hydrophilic coating and strong cation exchange functionality provide good mass transfer characteristics for a range of solute sizes.
  • Optimal design for effective separation of biomolecules
  • Pore sizes allow use of a range of solute sizes
  • Exceptional stability for long column lifetime
This process is controlled to provide the optimum density of strong cation exchange moieties for the analysis, separation and purification of a wide range of biomolecules from small peptides to large proteins. The 5 µm media delivers separations at higher resolution with the 30 µm media used for medium pressure liquid chromatography.
RENDELÉS
Többet mutat / Kevesebbet mutat
Kolonna formája Pórusméret Részecskeméret Belső átmérő × hossz M.egys VWR katalógus szám Mennyiség
Analytical 1000 Å 5 µm 2,1×50 mm 1 VARIPL1945-1502
Analytical 1000 Å 5 µm 4,6×50 mm 1 VARIPL1545-1502
Analytical 1000 Å 8 µm 2,1×50 mm 1 VARIPL1945-1802
Analytical 1000 Å 8 µm 2,1×150 mm 1 VARIPL1945-3802
Analytical 1000 Å 8 µm 4,6×50 mm 1 VARIPL1545-1802
Analytical 1000 Å 8 µm 4,6×150 mm 1 VARIPL1545-3802
Analytical 1000 Å 8 µm 7,5×50 mm 1 VARIPL1145-1802
Analytical 1000 Å 8 µm 7,5×150 mm 1 VARIPL1145-3802
Analytical 1000 Å 10 µm 4,6×150 mm 1 VARIPL1545-3102
Analytical 1000 Å 10 µm 4,6×250 mm 1 VARIPL1545-5102
Analytical 1000 Å 30 µm 4,6×150 mm 1 VARIPL1545-3702
Analytical 1000 Å 30 µm 4,6×250 mm 1 VARIPL1545-5702
Analytical 4000 Å 5 µm 2,1×50 mm 1 VARIPL1945-1503
Analytical 4000 Å 5 µm 4,6×50 mm 1 VARIPL1545-1503
Analytical 4000 Å 8 µm 2,1×50 mm 1 VARIPL1945-1803
Analytical 4000 Å 8 µm 2,1×150 mm 1 VARIPL1945-3803
Analytical 4000 Å 8 µm 4,6×50 mm 1 VARIPL1545-1803
Analytical 4000 Å 8 µm 4,6×150 mm 1 VARIPL1545-3803
Analytical 4000 Å 8 µm 7,5×50 mm 1 VARIPL1145-1803
Analytical 4000 Å 10 µm 4,6×150 mm 1 VARIPL1545-3103
Analytical 4000 Å 10 µm 4,6×250 mm 1 VARIPL1545-5103
Analytical 4000 Å 30 µm 4,6×150 mm 1 VARIPL1545-3703
Analytical 4000 Å 30 µm 4,6×250 mm 1 VARIPL1545-5703
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség