Szeretne újra bejelentkezni?
Strong anion exchange HPLC columns, PL-SAX
Gyártó: Agilent Technologies
These columns are ideal for the anion exchange HPLC separations of proteins and deprotected synthetic oligonucleotides under denaturing conditions.
  • The strong anion exchange functionality and covalently linked to a chemically stable polymer that extends the operating pH range
  • Wide range of particle sizes for flexible analysis to scale-up purification
  • Small particles deliver excellent chromatographic performance
  • The anion exchange capacity is independent of pH
  • Exceptional stability for long column lifetime
For synthetic oligonucleotides, separations using denaturing conditions of temperature, organic solvent, and high pH are all possible. It delivers improved chromatography for self-complementary or G-rich sequences that may associate to form aggregates or hairpin structures. The 5 µm material provides high efficiency separations of n and n-1 sequences.
A wide range of particle sizes and column geometries permits analysis and scale-up to purification. The strong anion exchange functionality provides a material with exceptional chemical and thermal stability, even with sodium hydroxide eluents, leading to long column lifetime.
RENDELÉS
Többet mutat / Kevesebbet mutat
Kolonna formája Fázis Pórusméret Részecskeméret Belső átmérő × hossz M.egys VWR katalógus szám Mennyiség
Analytical PL-SAX 1000 Å 5 µm 2,1×50 mm 1 VARIPL1951-1502
Analytical PL-SAX 1000 Å 5 µm 4,6×50 mm 1 VARIPL1551-1502
Analytical PL-SAX 1000 Å 8 µm 2,1×50 mm 1 VARIPL1951-1802
Analytical PL-SAX 1000 Å 8 µm 2,1×150 mm 1 VARIPL1951-3802
Analytical PL-SAX 1000 Å 8 µm 4,6×150 mm 1 VARIPL1551-3802
Analytical PL-SAX 1000 Å 8 µm 4,6×50 mm 1 VARIPL1551-1802
Analytical PL-SAX 1000 Å 8 µm 7,5×50 mm 1 VARIPL1151-1802
Analytical PL-SAX 1000 Å 8 µm 7,5×150 mm 1 VARIPL1151-3802
Analytical PL-SAX 1000 Å 10 µm 4,6×150 mm 1 VARIPL1551-3102
Analytical PL-SAX 1000 Å 10 µm 4,6×250 mm 1 VARIPL1551-5102
Analytical PL-SAX 1000 Å 30 µm 4,6×150 mm 1 VARIPL1551-3702
Analytical PL-SAX 1000 Å 30 µm 4,6×250 mm 1 VARIPL1551-5702
Analytical PL-SAX 4000 Å 5 µm 2,1×50 mm 1 VARIPL1951-1503
Analytical PL-SAX 4000 Å 5 µm 4,6×50 mm 1 VARIPL1551-1503
Analytical PL-SAX 4000 Å 8 µm 2,1×50 mm 1 VARIPL1951-1803
Analytical PL-SAX 4000 Å 8 µm 2,1×150 mm 1 VARIPL1951-3803
Analytical PL-SAX 4000 Å 8 µm 4,6×50 mm 1 VARIPL1551-1803
Analytical PL-SAX 4000 Å 8 µm 4,6×150 mm 1 VARIPL1551-3803
Analytical PL-SAX 4000 Å 8 µm 7,5×50 mm 1 VARIPL1151-1803
Analytical PL-SAX 4000 Å 8 µm 7,5×150 mm 1 VARIPL1151-3803
Analytical PL-SAX 4000 Å 10 µm 4,6×150 mm 1 VARIPL1551-3103
Analytical PL-SAX 4000 Å 10 µm 4,6×250 mm 1 VARIPL1551-5103
Analytical PL-SAX 4000 Å 30 µm 4,6×150 mm 1 VARIPL1551-3703
Analytical PL-SAX 4000 Å 30 µm 4,6×250 mm 1 VARIPL1551-5703
Vonalkód generálása Barcode Label Format: 
PDF katalógus létrehozása
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Vagy
Review & Compare Alternatives
Original Product:
Leírás Katalógusszám Availability Egység Az Ön ára Price Per Mennyiség